Learn More
Diethyl 2-Acetamido-2-[2-(4-octylphenyl)ethyl]propanedioate, Mikromol
Description
Diethyl 2-Acetamido-2-[2-(4-octylphenyl)ethyl]propanedioate, Mikromol
Specifications
Specifications
| Chemical Name or Material | Diethyl 2-Acetamido-2-[2-(4-octylphenylethyl)malonate |
| CAS | 162358-08-9 |
| Analyte or Component Names | Diethyl 2-Acetamido-2-[2-(4-octylphenylethyl)malonate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H39 N O5 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Propanedioic acid, 2-(acetylamino)-2-[2-(4-octylphenyl)ethyl]-, 1,3-diethyl ester, Propanedioic acid, (acetylamino)[2-(4-octylphenyl)ethyl]-, diethyl ester (9CI), (Acetylamino)[2-(4-octylphenyl)ethyl]propanedioic acid diethyl ester, 2-Acetylamino-2-(2-(4-octylphenyl)ethyl)propanedioic acid diethyl ester, Diethyl 2-(acetamido)-2-(2-(4-octylphenyl)ethyl)propanedioate, Diethyl 2-[2-(4-n-octylphenyl)ethyl]-2-acetamidomalonate |
| InChI Formula | InChI=1S/C25H39NO5/c1-5-8-9-10-11-12-13-21-14-16-22(17-15-21)18-19-25(26-20(4)27,23(28)30-6-2)24(29)31-7-3/h14-17H,5-13,18-19H2,1-4H3,(H,26,27) |
| Solution Type | Neat |
| SMILES | CCCCCCCCc1ccc(CCC(NC(=O)C)(C(=O)OCC)C(=O)OCC)cc1 |
| Show More |