Learn More
Diclofenac Sodium EP Impurity C, Mikromol™
Product Code. 30462656
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462656 4 x 25 mg 100mg
17971156 25 mg 25mg
17954246 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30462656 Supplier Mikromol Supplier No. MM0006.0400254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Diclofenac Sodium EP Impurity C, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2-[(2,6-Dichlorophenyl)amino]phenyl)methanol
CAS 27204-57-5
Analyte or Component Names (2-[(2,6-Dichlorophenyl)amino]phenyl)methanol
Shipping Condition Room Temperature
Molecular Formula C13 H11 Cl2 N O
Certifications/Compliance ISO 17025
Synonym [2-[(2,6-Dichlorophenyl)amino]phenyl]methanol, Diclofenac Sodium Imp. C (EP), Diclofenac Imp. C (EP), [2-[(2,6-Dichlorophenyl)amino]phenyl]methanol, Diclofenac Sodium Impurity C, Diclofenac Impurity C
InChI Formula InChI=1S/C13H11Cl2NO/c14-10-5-3-6-11(15)13(10)16-12-7-2-1-4-9(12)8-17/h1-7,16-17H,8H2
Solution Type Neat
SMILES OCc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC Name [2-(2,6-dichloroanilino)phenyl]methanol
Molecular Weight (g/mol) 268.14
Quantity 4 x 25 mg
Formula Weight 267.0218 g/mol