Learn More
Dichromate Standard: Cr2O7(-2) @ 1000 μg/mL in H2O, VHG™
Product Code. 30003198
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 30003198
Brand: VHG Labs VHGIDCRO500
Please to purchase this item. Need a web account? Register with us today!
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Dichromate Standard: Cr2O7(-2) @ 1000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dichromate
CAS 13907-47-6
Methods Ion Chromatography
Analyte or Component Names Dichromate
Shipping Condition Room Temperature
Molecular Formula (Cr2 O7)2-
Synonym Cr2O72−
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
IUPAC Name Dichromate
Solvent or Matrix Water
Molecular Weight (g/mol) 215.99
Quantity 500 mL
Formula Weight 215.99 g/mol