missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II), Pd 13% min

Product Code. p-7059603
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15476655

Brand: Thermo Scientific Alfa Aesar 044977.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 205319-06-8 | C36H28Cl2OP2Pd | 715.89 g/mol

Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II) is used as a catalyst for cross-coupling reaction of 2-brom-1,3-dienes to prepare high stereoselectivity of conjugated Z,E dienes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205319-06-8
Molecular Formula C36H28Cl2OP2Pd
Molecular Weight (g/mol) 715.89
MDL Number MFCD09953446
InChI Key QZIOEBWDZNUTLZ-UHFFFAOYSA-L
Synonym dichloro bis diphenylphosphinophenyl ether palladium ii, dichloro bis 2-diphenylphosphino phenyl ether palladium ii, bis diphenylphosphinophenyl ether palladium ii dichloride, dichloro bis diphenylphosphinophenyl ether palladium ii , pd, oxydi 2,1-phenylene bis diphenylphosphane-dichloropalladium 1/1, palladium chloride; 2-2-diphenylphosphanyl phenoxy phenyl diphenylphosphane
PubChem CID 11377567
IUPAC Name dichloropalladium;[2-(2-diphenylphosphanylphenoxy)phenyl]-diphenylphosphane
SMILES Cl[Pd++]Cl.O(C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 715.88
Concentration or Composition (by Analyte or Components) Pd 14.8%
Chemical Name or Material Dichloro[bis (diphenylphosphinophenyl)ether]palladium (II)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.