accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 205319-06-8

IUPAC Name: dichloropalladium; {2-[2-(diphenylphosphanyl)phenoxy]phenyl}diphenylphosphane
Synonyms: palladium chloride; {2-[2-(diphenylphosphanyl)phenoxy]phenyl}diphenylphosphane
Molecular Weight (g/mol): 715.89
Molecular Formula: C36H28Cl2OP2Pd
InChi Key: VYOUBMJFTDMKRR-UHFFFAOYSA-L
SMILES: Cl[Pd]Cl.O(C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results