Learn More
Di-n-Hexyl Phthalate, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Di-n-hexyl phthalate | 84-75-3 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Packaging | 2mL amber prescored glass ampule |
| CAS | 84-75-3 |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| Molecular Formula | C20H30O4 |
| MDL Number | MFCD00043720 |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| InChI Key | KCXZNSGUUQJJTR-UHFFFAOYSA-N |
| SMILES | CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC |
| IUPAC Name | 1,2-dihexyl benzene-1,2-dicarboxylate |
| Solvent or Matrix | Hexane |
| Show More |
Product intended for laboratory use only