Learn More
Dexchlorpheniramine N-Oxide Dihydrochloride, Mikromol™
Description
Dexchlorpheniramine N-Oxide Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | Dexchlorpheniramine N-Oxide Dihydrochloride |
| CAS | 2725579-30-4 |
| Analyte or Component Names | Dexchlorpheniramine N-Oxide Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H19 Cl N2 O . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Dexchlorpheniramine N-Oxide Dihydrochloride |
| InChI Formula | InChI=1S/C16H19ClN2O.2ClH/c1-19(2,20)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;;/h3-9,11,15H,10,12H2,1-2H3;2*1H/t15-;;/m0../s1 |
| SMILES | Cl.Cl.C[N+](C)([O-])CC[C@@H](c1ccc(Cl)cc1)c2ccccn2 |
| IUPAC Name | (3S)-3-(4-chlorophenyl)-N,N-dimethyl-3-pyridin-2-ylpropan-1-amine oxide;dihydrochloride |
| Show More |