Learn More
Desamino 3’’-Azido Tetra-N-Boc Tobramycin 6’’-Carbamate O-Tetracetate, TRC
Product Code. 30355051
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355051 1 mg 1mg
30350247 5 mg 5mg
30366569 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30355051 Supplier Toronto Research Chemicals Supplier No. TRCD2975901MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Desamino 3''-Azido Tetra-N-Boc Tobramycin 6''-Carbamate O-Tetracetate
Analyte or Component Names Desamino 3''-Azido Tetra-N-Boc Tobramycin 6''-Carbamate O-Tetracetate
Shipping Condition Room Temperature
Molecular Formula C47H76N8O22
InChI Formula InChI=1S/C47H76N8O22/c1-21(56)66-28-18-27(53-43(64)77-47(14,15)16)37(70-29(28)19-50-40(61)74-44(5,6)7)72-32-25(51-41(62)75-45(8,9)10)17-26(52-42(63)76-46(11,12)13)33(36(32)69-24(4)59)73-38-35(68-23(3)58)31(54-55-49)34(67-22(2)57)30(71-38)20-65-39(48)60/h25-38H,17-20H2,1-16H3,(H2,48,60)(H,50,61)(H,51,62)(H,52,63)(H,53,64)/t25?,26?,27?,28?,29-,30-,31?,32-,33+,34?,35?,36?,37-,38-/m1/s1
Solution Type Neat
SMILES NC(OC[C@H]1O[C@H](O[C@H]2[C@H](NC(OC(C)(C)C)=O)C[C@H](NC(OC(C)(C)C)=O)[C@@H](O[C@@H]3[C@H](NC(OC(C)(C)C)=O)C[C@H](OC(C)=O)[C@@H](CNC(OC(C)(C)C)=O)O3)[C@@H]2OC(C)=O)[C@H](OC(C)=O)[C@@H](N=[N+]=[N-])[C@@H]1OC(C)=O)=O
IUPAC Name (2S,3R,4S,5S,6R)-2-(((1S,2S,3R,4S,6R)-2-acetoxy-3-(((2R,3R,5S,6R)-5-acetoxy-3-((tert-butoxycarbonyl)amino)-6-(((tert-butoxycarbonyl)amino)methyl)tetrahydro-2H-pyran-2-yl)oxy)-4,6-bis((tert-butoxycarbonyl)amino)cyclohexyl)oxy)-4-azido-6-((carbamoyloxy)methyl)tetrahydro-2H-pyran-3,5-diyl diacetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 1105.15
Quantity 1 mg
Formula Weight 1104.51