Learn More
Deacetyl Racecadotril Disulfide, TRC
Product Code. 30368082
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368082 500 mg 500mg
30357326 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30368082 Supplier Toronto Research Chemicals Supplier No. TRCD199020500MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 141437-88-9
Molecular Formula C38H40N2O6S2
Molecular Weight (g/mol) 684.86
Synonym N,N'-[Dithiobis[1-oxo-2-(phenylmethyl)-3,1-propanediyl]]bis-glycine Bis(phenylmethyl) Ester
IUPAC Name benzyl 2-[[2-benzyl-3-[[2-benzyl-3-oxo-3-[(2-oxo-2-phenylmethoxyethyl)amino]propyl]disulfanyl]propanoyl]amino]acetate
SMILES O=C(NCC(OCC1=CC=CC=C1)=O)C(CSSCC(C(NCC(OCC2=CC=CC=C2)=O)=O)CC3=CC=CC=C3)CC4=CC=CC=C4
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C38H40N2O6S2/c41-35(45-25-31-17-9-3-10-18-31)23-39-37(43)33(21-29-13-5-1-6-14-29)27-47-48-28-34(22-30-15-7-2-8-16-30)38(44)40-24-36(42)46-26-32-19-11-4-12-20-32/h1-20,33-34H,21-28H2,(H,39,43)(H,40,44)
Chemical Name or Material Deacetyl Racecadotril Disulfide