CAS RN 141437-88-9
CAS RN 141437-88-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Deacetyl Racecadotril Disulfide, TRC
CAS: 141437-88-9 Molecular Formula: C38H40N2O6S2 Molecular Weight (g/mol): 684.86 Synonym: N,N'-[Dithiobis[1-oxo-2-(phenylmethyl)-3,1-propanediyl]]bis-glycine Bis(phenylmethyl) Ester IUPAC Name: benzyl 2-[[2-benzyl-3-[[2-benzyl-3-oxo-3-[(2-oxo-2-phenylmethoxyethyl)amino]propyl]disulfanyl]propanoyl]amino]acetate SMILES: O=C(NCC(OCC1=CC=CC=C1)=O)C(CSSCC(C(NCC(OCC2=CC=CC=C2)=O)=O)CC3=CC=CC=C3)CC4=CC=CC=C4