Learn More
Dantrolene USP Related Compound C, Mikromol™
Product Code. 30052655
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052655 25 mg 25mg
17963196 100 mg 100mg
30464214 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30052655 Supplier Mikromol Supplier No. MM0905.010025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Dantrolene USP Related Compound C
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-(4-nitrophenyl)-2-furancarboxyaldehyde
CAS 7147-77-5
Analyte or Component Names 5-(4-nitrophenyl)-2-furancarboxyaldehyde
Shipping Condition Room Temperature
Molecular Formula C11 H7 N O4
Certifications/Compliance ISO 17025
Synonym 5-(4-Nitrophenyl)furan-2-carbaldehyde, (5-(4-nitrophenyl)-2-furancarboxyaldehyde, Dantrolene Related Compound C
InChI Formula InChI=1S/C11H7NO4/c13-7-10-5-6-11(16-10)8-1-3-9(4-2-8)12(14)15/h1-7H
SMILES [O-][N+](=O)c1ccc(cc1)c2oc(C=O)cc2
IUPAC Name 5-(4-nitrophenyl)furan-2-carbaldehyde
Molecular Weight (g/mol) 217.18
Quantity 25 mg
Formula Weight 217.0375 g/mol