missing translation for 'onlineSavingsMsg'
Learn More

Dantrolene USP Related Compound C, Mikromol™

Product Code. 17963196
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963196 100 mg 100mg
30052655 25 mg 25mg
30464214 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17963196 Supplier Mikromol Supplier No. MM0905.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dantrolene USP Related Compound C
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-(4-nitrophenyl)-2-furancarboxyaldehyde
CAS 7147-77-5
Analyte or Component Names 5-(4-nitrophenyl)-2-furancarboxyaldehyde
Shipping Condition Room Temperature
Molecular Formula C11 H7 N O4
Certifications/Compliance ISO 17025
Synonym 5-(4-Nitrophenyl)furan-2-carbaldehyde, (5-(4-nitrophenyl)-2-furancarboxyaldehyde, Dantrolene Related Compound C
InChI Formula InChI=1S/C11H7NO4/c13-7-10-5-6-11(16-10)8-1-3-9(4-2-8)12(14)15/h1-7H
SMILES [O-][N+](=O)c1ccc(cc1)c2oc(C=O)cc2
IUPAC Name 5-(4-nitrophenyl)furan-2-carbaldehyde
Molecular Weight (g/mol) 217.18
Quantity 100 mg
Formula Weight 217.0375 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.