Learn More
Memmert™ DAkkS Factory Calibration for UN110
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN110 Oven
Unit Size:
Each
Description
- For one free-selectable temperature value according to method C (DKD-R 5-7)
Specifications
Specifications
| For Use With (Equipment) | UN110 Oven |
| Product Type | DAkkS Factory Calibration |