Learn More
Memmert™ DAkkS Factory Calibration for IN110
Product Code. 17557365 Shop All Memmert Products
Click to view available options
Unit Size:
Each
Product Code. 17557365
Brand: Memmert™ IN110E39696
Please to purchase this item. Need a web account? Register with us today!
DAkkS factory calibration for a freely selectable temperature for Memmert Incubators.
  • For one free-selectable temperature value according to method C (DKD-R 5-7)
Specifications
For Use With (Equipment) IN110 Incubator
Product Type DAkkS Factory Calibration