missing translation for 'onlineSavingsMsg'
Learn More

D-Tyrosine ethyl ester hydrochloride, 98%, Thermo Scientific™

Product Code. 15408748
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408748 5 g 5g
15418748 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408748 Supplier Thermo Scientific Alfa Aesar Supplier No. H62194.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23234-43-7
Molecular Formula C11H16ClNO3
Molecular Weight (g/mol) 245.70
MDL Number MFCD00180750
InChI Key BQULAXAVRFIAHN-OXZMBGLCNA-N
Synonym h-d-tyr-oet.hcl, h-d-tyr-oet hcl, d-tyrosine ethyl ester hydrochloride, r-ethyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, ethyl d-tyrosinate hcl, ethyl d-tyrosinate hydrochloride, ethyl 2r-2-amino-3-4-hydroxyphenyl propanoate hydrochloride, h-d-tyr-oethcl, h-d-tyr-oet?cl, h-d-tyr-oet. hcl
PubChem CID 45357981
IUPAC Name ethyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride
SMILES Cl.CCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1

Specifications

Quantity 5 g
Formula Weight 245.7
Percent Purity 98%
Chemical Name or Material D-Tyrosine ethyl ester hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.