accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HONH2(R)OOHClCH3

CAS RN 23234-43-7

IUPAC Name: ethyl 2-amino-3-(4-hydroxyphenyl)propanoate hydrochloride
Synonyms: L-tyrosine, ethyl ester hydrochloride
Molecular Weight (g/mol): 245.7
Molecular Formula: C11H16ClNO3
InChi Key: BQULAXAVRFIAHN-UHFFFAOYNA-N
SMILES: Cl.CCOC(=O)C(N)CC1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results