Learn More
Thermo Scientific™ [D-Phe12, Nle21,38, Glu30, Lys33] Corticotropin Releasing Factor (12-41), Human
186.70 € - 340.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15475805
|
Thermo Scientific™
J66247.!D |
0.5 mg |
248.00 €
0.50mg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15474475
|
Thermo Scientific™
J66247.!D |
0.5 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15484475
|
Thermo Scientific™
J66247.M^ |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Format: Solid
Specifications
Store at -20°C | |
Solid |