missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ [D-Phe12, Nle21,38, Glu30, Lys33] Corticotropin Releasing Factor (12-41), Human

Brand:  Thermo Scientific™ J66247.|D

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15474475

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Format: Solid
Specifications

Specifications

Store at -20°C
Solid
Corticotropin Releasing Factor (12-41)
0.5 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions