Learn More
D,L-Mevalonic Acid-d3 Dicyclohexylamine Salt, TRC
Product Code. 30133458
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133458 10 mg 10mg
17918509 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30133458 Supplier Toronto Research Chemicals Supplier No. TRCM33902210MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1329796-98-6
Molecular Formula C18H32D3NO4
Molecular Weight (g/mol) 332.49
Synonym 3,5-Dihydroxy-3-(methyl-d3) Pentanoic Acid Dicyclohexylamine Salt, Hiochic Acid-d3 Dicyclohexylamine Salt, 3,5-Dihydroxy-3-(methyl-d3)valeric Acid Dicyclohexylamine Salt, 2,4-Dideoxy-3-C-(methyl-d3)
IUPAC Name dicyclohexylamine 3,5-dihydroxy-3-(methyl-d3)pentanoate
SMILES OCCC(O)(C([2H])([2H])[2H])CC(O)=O.C1(CCCCC1)NC2CCCCC2
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C12H23N.C6H12O4/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-6(10,2-3-7)4-5(8)9/h11-13H,1-10H2;7,10H,2-4H2,1H3,(H,8,9)/i;1D3
Formula Weight 332.28
Chemical Name or Material D,L-Mevalonic Acid-d3 Dicyclohexylamine Salt