CAS RN 1329796-98-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
D,L-Mevalonic Acid-d3 Dicyclohexylamine Salt, TRC
CAS: 1329796-98-6 Molecular Formula: C18H32D3NO4 Molecular Weight (g/mol): 332.49 Synonym: 3,5-Dihydroxy-3-(methyl-d3) Pentanoic Acid Dicyclohexylamine Salt,Hiochic Acid-d3 Dicyclohexylamine Salt,3,5-Dihydroxy-3-(methyl-d3)valeric Acid Dicyclohexylamine Salt,2,4-Dideoxy-3-C-(methyl-d3) IUPAC Name: dicyclohexylamine 3,5-dihydroxy-3-(methyl-d3)pentanoate SMILES: OCCC(O)(C([2H])([2H])[2H])CC(O)=O.C1(CCCCC1)NC2CCCCC2