Learn More
D-Alanine-3,3,3-d3, TRC
Product Code. 30179163
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179163 100 mg 100mg
30179526 50 mg 50mg
30179080 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179163 Supplier Toronto Research Chemicals Supplier No. TRCA480998100MG
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 177614-69-6
Molecular Formula C3 D3 H4 N O2
Molecular Weight (g/mol) 92.1117
Synonym D-Alanine-3,3,3-d3 (9CI, ACI)
IUPAC Name (2R)-2-amino-3,3,3-trideuteriopropanoic acid
SMILES [2H]C([2H])([2H])[C@@H](N)C(=O)O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m1/s1/i1D3
Formula Weight 92.0665
Chemical Name or Material D-Alanine-3,3,3-d3