CAS RN 177614-69-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
D-Alanine-3,3,3-d3, TRC
CAS: 177614-69-6 Molecular Formula: C3 D3 H4 N O2 Molecular Weight (g/mol): 92.1117 Synonym: D-Alanine-3,3,3-d3 (9CI, ACI) IUPAC Name: (2R)-2-amino-3,3,3-trideuteriopropanoic acid SMILES: [2H]C([2H])([2H])[C@@H](N)C(=O)O
2
D-Alanine-3,3,3-d3, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.