missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP46A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
213.00 € - 450.00 €
Specifications
| Antigen | CYP46A1 |
|---|---|
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18270550
|
Novus Biologicals
NB400-140SS |
0.025 mL |
213.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18775063
|
Novus Biologicals
NB400-140 |
0.1 mL |
450.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CYP46A1 Polyclonal specifically detects CYP46A1 in Human, Rabbit, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CYP46A1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism, Neuroscience | |
| Whole antisera with No Preservative | |
| CH24H, cholesterol 24-hydroxylase, CYP46CP46, Cytochrome P450 46A1, cytochrome P450, family 46, subfamily A, polypeptide 1, cytochrome P450, subfamily 46 (cholesterol 24-hydroxylase), EC 1.14.13.98 | |
| CYP46A1 | |
| IgG | |
| Unpurified | |
| Human and Rabbit. Does not react with mouse. |
| Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:200 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Rabbit, Mouse (Negative) | |
| Q9Y6A2 | |
| 10858 | |
| A synthetic peptide corresponding to residues between 200-250 of human CYP46A1. [UniProt# Q9Y6A2] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title