missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP46A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB400-140
This item is not returnable.
View return policy
Description
CYP46A1 Polyclonal specifically detects CYP46A1 in Human, Rabbit, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CYP46A1 | |
| Polyclonal | |
| Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:200 | |
| Q9Y6A2 | |
| CYP46A1 | |
| A synthetic peptide corresponding to residues between 200-250 of human CYP46A1. [UniProt# Q9Y6A2] | |
| 0.1 mL | |
| Lipid and Metabolism, Neuroscience | |
| 10858 | |
| Human, Rabbit, Mouse (Negative) | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with No Preservative | |
| CH24H, cholesterol 24-hydroxylase, CYP46CP46, Cytochrome P450 46A1, cytochrome P450, family 46, subfamily A, polypeptide 1, cytochrome P450, subfamily 46 (cholesterol 24-hydroxylase), EC 1.14.13.98 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Human and Rabbit. Does not react with mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction