Learn More
Cyclopentolate N-Oxide Hydrochloride, Mikromol™
Description
Cyclopentolate N-Oxide Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Cyclopentolate N-Oxide Hydrochloride |
| CAS | 2724726-92-3 |
| Analyte or Component Names | Cyclopentolate N-Oxide Hydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C17 H25 N O4 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Cyclopentolate N-Oxide Hydrochloride |
| InChI Formula | InChI=1S/C17H25NO4.ClH/c1-18(2,21)12-13-22-16(19)15(14-8-4-3-5-9-14)17(20)10-6-7-11-17;/h3-5,8-9,15,20H,6-7,10-13H2,1-2H3;1H |
| SMILES | Cl.C[N+](C)([O-])CCOC(=O)C(c1ccccc1)C2(O)CCCC2 |
| IUPAC Name | 2-[2-(1-hydroxycyclopentyl)-2-phenylacetyl]oxy-N,N-dimethylethanamine oxide;hydrochloride |
| Show More |