missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyldiphenylphosphine, 98%

Product Code. p-96735 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10278903

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6372-42-5 | C18H21P | 268.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6372-42-5
Molecular Formula C18H21P
Molecular Weight (g/mol) 268.34
MDL Number MFCD00046360
InChI Key ZXKWUYWWVSKKQZ-UHFFFAOYSA-N
Synonym cyclohexyldiphenylphosphine, phosphine, cyclohexyldiphenyl, cyclohexyl diphenyl phosphine, diphenylcyclohexylphosphine, diphenylphosphinocyclohexane, cyclohexyl-diphenyl-phosphane, pubchem6451, acmc-1ba4g, cyclohexyl diphenyl phosphane
PubChem CID 80756
IUPAC Name cyclohexyl(diphenyl)phosphane
SMILES C1CCC(CC1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 53.0°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 268.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Cyclohexyldiphenylphosphine
Show More Show Less
compliance-icons
Product Identifier
  • Cyclohexyldiphenylphosphine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.