accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6372-42-5

IUPAC Name: cyclohexyldiphenylphosphane
Synonyms: cyclohexyldiphenylphosphane
Molecular Weight (g/mol): 268.34
Molecular Formula: C18H21P
InChi Key: ZXKWUYWWVSKKQZ-UHFFFAOYSA-N
SMILES: C1CCC(CC1)P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results