Learn More
Cycloguanil Hydrochloride, Mikromol™
Description
Cycloguanil Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Cycloguanil Hydrochloride |
| CAS | 152-53-4 |
| Analyte or Component Names | Cycloguanil Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H14 Cl N5 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1,3,5-Triazine-2,4-diamine, 1-(4-chlorophenyl)-1,6-dihydro-6,6-dimethyl-, hydrochloride (1:1)1,3,5-Triazine-2,4-diamine, 1-(4-chlorophenyl)-1,6-dihydro-6,6-dimethyl-, monohydrochloride (9CI), s-Triazine, 4,6-diamino-1-(p-chlorophenyl)-1,2-dihydro-2,2-dimethyl-, hydrochloride (6CI), s-Triazine, 4,6-diamino-1-(p-chlorophenyl)-1,2-dihydro-2,2-dimethyl-, monohydrochloride (8CI), 1-(4-Chlorophenyl)-1,6-dihydro-6,6-dimethyl-1,3,5-triazine-2,4-diamine monohydrochloride, BN 2410, Chlorcycloguanil, Chloroguanide triazine hydrochloride, Cycloguanil hydrochloride, NSC 3074 |
| InChI Formula | InChI=1S/C11H14ClN5.ClH/c1-11(2)16-9(13)15-10(14)17(11)8-5-3-7(12)4-6-8;/h3-6H,1-2H3,(H4,13,14,15,16);1H |
| SMILES | Cl.CC1(C)N=C(N)N=C(N)N1c2ccc(Cl)cc2 |
| IUPAC Name | 1-(4-chlorophenyl)-6,6-dimethyl-1,3,5-triazine-2,4-diamine;hydrochloride |
| Show More |