Learn More
Cycloguanil Hydrochloride, LoGiCal
Product Code. 30463669
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463669 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30463669 Supplier LoGiCal Supplier No. LGCFOR1580.00
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Cycloguanil Hydrochloride
CAS 152-53-4
Analyte or Component Names Cycloguanil Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C11 H14 Cl N5 . Cl H
Certifications/Compliance ISO 9001
Synonym 1,3,5-Triazine-2,4-diamine, 1-(4-chlorophenyl)-1,6-dihydro-6,6-dimethyl-, hydrochloride (1:1)1,3,5-Triazine-2,4-diamine, 1-(4-chlorophenyl)-1,6-dihydro-6,6-dimethyl-, monohydrochloride (9CI), s-Triazine, 4,6-diamino-1-(p-chlorophenyl)-1,2-dihydro-2,2-dimethyl-, hydrochloride (6CI), s-Triazine, 4,6-diamino-1-(p-chlorophenyl)-1,2-dihydro-2,2-dimethyl-, monohydrochloride (8CI), 1-(4-Chlorophenyl)-1,6-dihydro-6,6-dimethyl-1,3,5-triazine-2,4-diamine monohydrochloride, BN 2410, Chlorcycloguanil, Chloroguanide triazine hydrochloride, Cycloguanil hydrochloride, NSC 3074
InChI Formula InChI=1S/C11H14ClN5.ClH/c1-11(2)16-9(13)15-10(14)17(11)8-5-3-7(12)4-6-8;/h3-6H,1-2H3,(H4,13,14,15,16);1H
Solution Type Neat
SMILES Cl.CC1(C)N=C(N)N=C(N)N1c2ccc(Cl)cc2
IUPAC Name 1-(4-chlorophenyl)-6,6-dimethyl-1,3,5-triazine-2,4-diamine;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 288.18
Quantity 10 mg
Formula Weight 287.0705 g/mol