Learn More
Cyclanilide, TRC
Product Code. 30367038
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367038 500 mg 500mg
30351901 250 mg 250mg
30366395 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30367038 Supplier Toronto Research Chemicals Supplier No. TRCC987765500MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 113136-77-9
Molecular Formula C11 H9 Cl2 N O3
Molecular Weight (g/mol) 274.1
Synonym 1-[[(2,4-Dichlorophenyl)amino]carbonyl]cyclopropanecarboxylic acid, Cyclanilide, RPA 90946
IUPAC Name 1-[(2,4-dichlorophenyl)carbamoyl]cyclopropane-1-carboxylic acid
SMILES OC(=O)C1(CC1)C(=O)Nc2ccc(Cl)cc2Cl
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C11H9Cl2NO3/c12-6-1-2-8(7(13)5-6)14-9(15)11(3-4-11)10(16)17/h1-2,5H,3-4H2,(H,14,15)(H,16,17)
Formula Weight 272.9959
Chemical Name or Material Cyclanilide