CAS RN 113136-77-9
CAS RN 113136-77-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cyclanilide, TRC
CAS: 113136-77-9 Molecular Formula: C11 H9 Cl2 N O3 Molecular Weight (g/mol): 274.1 Synonym: 1-[[(2,4-Dichlorophenyl)amino]carbonyl]cyclopropanecarboxylic acid,Cyclanilide,RPA 90946 IUPAC Name: 1-[(2,4-dichlorophenyl)carbamoyl]cyclopropane-1-carboxylic acid SMILES: OC(=O)C1(CC1)C(=O)Nc2ccc(Cl)cc2Cl