Learn More
CST4 Monoclonal Antibody (OTI2C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504095S
Description
CST4 Monoclonal antibody specifically detects CST4 in Human samples. It is validated for Western BlotSpecifications
| CST4 | |
| Monoclonal | |
| 0.91 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WWV3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84816 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2C2 | |
| Unconjugated | |
| Rtn4ip1 | |
| CST4; cystatin 4; cystatin S; cystatin-4; Cystatin-S; cystatin-SA-III; Salivary acidic protein 1 | |
| Human recombinant protein fragment corresponding to amino acids 41-279 of human RTN4IP1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.