Learn More
CST4 Monoclonal Antibody (OTI2C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504095
Description
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions. The cystatin locus on chromosome 20 contains the majority of the type 2 cystatin genes and pseudogenes. This gene is located in the cystatin locus and encodes a type 2 salivary cysteine peptidase inhibitor. The protein is an S-type cystatin, based on its high level of expression in saliva, tears and seminal plasma. The specific role in these fluids is unclear but antibacterial and antiviral activity is present, consistent with a protective function.Specifications
| CST4 | |
| Monoclonal | |
| 0.91 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P01036 | |
| CST4 | |
| Full length human recombit protein of human CST4 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2C2 | |
| Unconjugated | |
| CST4 | |
| cystatin S | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1472 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.