missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cresol Red sodium salt

Product Code. 11339719
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339719 100 g 100g
11329719 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339719 Supplier Thermo Scientific Chemicals Supplier No. B21361.22
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

Transition interval (acid): pH 0.2 (red) to pH 1.8 (yellow); (alkaline) pH 7.2 (yellow) to pH 8.8 (red) | CAS: 62625-29-0 | C21H17NaO5S

Cresol Red sodium salt acts as a constituent of polymerase chain reaction reagent systems. It is useful as a pH indicator with color changes with pH 0.2 - pH 1.8 and pH 7.1 - 8.8. Further, it plays an important role in molecular biology reactions in place of other loading dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cresol Red sodium salt acts as a constituent of polymerase chain reaction reagent systems. It is useful as a pH indicator with color changes with pH 0.2 - pH 1.8 and pH 7.1 - 8.8. Further, it plays an important role in molecular biology reactions in place of other loading dyes.

Solubility
Soluble in water, dilute alcohols, acids, alkalies and ethanol.

Notes
Transition interval (acid): pH 0.2 (red) to pH 1.8 (yellow); (alkaline) pH 7.2 (yellow) to pH 8.8 (red)Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-29-0
MDL Number MFCD00001618
Synonym o-Cresolsulfonephthalein sodium salt

Specifications

Melting Point ∼250°C (decomposition)
Molecular Formula C21H17NaO5S
Solubility Information Soluble in water,dilute alcohols,acids,alkalies and ethanol.
Formula Weight 404.42
Quantity 100 g
Chemical Name or Material Cresol Red sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.