accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NaH3COHOSOOOH3C

CAS RN 62625-29-0

IUPAC Name: sodium 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-2-methylbenzen-1-olate
Synonyms: sodium 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1λ⁶-benzoxathiol-3-yl]-2-methylbenzenolate
Molecular Weight (g/mol): 404.41
Molecular Formula: C21H17NaO5S
InChi Key: FMKJVYROEYMYFV-UHFFFAOYNA-M
SMILES: [Na+].CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C([O-])C(C)=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)