Learn More
Cortisone Acetate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Cortisone Acetate |
| CAS | 50-04-4 |
| Analyte or Component Names | Cortisone Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H30 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Hydrocortisone Acetate Imp. D (EP), 17-Hydroxy-3,11,20-Trioxo-pregn-4-en-21-yl Acetate, Cortisone Acetate, Hydrocortisone Acetate Imp. D (Pharmeuropa), Cortisone 21-acetate, 17,21-Dihydroxypregn-4-ene-3,11,20-trione 21 acetate, 21-Acetoxy-17-hydroxypregn-4-ene-3,11,20-trione, 21-Acetoxy-17alpha-hydroxy-3,11,20-triketopregnene-4, 21-Acetoxy-17alpha-hydroxypregn-4-ene-3,11,20-trione, 4-Pregnene-17a,21-diol-3,11,20-trione 21-acetate, Adreson, Artriona, Biocort Acetate, Compound E Acetate, Corlin, Cortadren, Cortelan, Cortisone acetate, Cortistab, Cortisyl, Cortisyl Artriona, Cortone acetate, Incortin, Irisone acetate, NSC 49420, Ricortex, Scheroson, Hydrocortisone Acetate Impurity D |
| InChI Formula | InChI=1S/C23H30O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h10,16-17,20,28H,4-9,11-12H2,1-3H3/t16-,17-,20+,21-,22-,23-/m0/s1 |
| Solution Type | Neat |
| SMILES | CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C |
| Show More |