missing translation for 'onlineSavingsMsg'
Learn More

Copper germanium selenide, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. p-7059913
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15462407 10 g 10g
15472407 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462407 Supplier Thermo Scientific Alfa Aesar Supplier No. 045605.09
Add to basket
This item is not returnable. View return policy

The most notable physical characteristics of germania (GeO2) are its high index of refraction and its low optical dispersion. These make it especially useful for wide-angle camera lenses, microscopy, and for the core part of optical fibers. Germanium-on-insulator substrates are seen as a potential replacement for silicon on miniaturized chips.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54723-90-9
Molecular Formula CuGeSe
Molecular Weight (g/mol) 215.15
MDL Number MFCD12546179
InChI Key LQTVKWORTHHMFT-UHFFFAOYSA-N
Synonym copper germanium selenide
PubChem CID 121233948
IUPAC Name selanylidenegermylidenecopper
SMILES [Cu]=[Ge]=[Se]

Specifications

Physical Form Pieces
Quantity 10 g
UN Number UN3283
Solubility Information Insoluble in water.
Formula Weight 215.12
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Copper germanium selenide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.