accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 54723-90-9

CuGeSe

CAS RN 54723-90-9

IUPAC Name: cupriogermaneselone
Synonyms: cupriogermaneselone
Molecular Weight (g/mol): 215.15
Molecular Formula: CuGeSe
InChi Key: LQTVKWORTHHMFT-UHFFFAOYSA-N
SMILES: [Cu]=[Ge]=[Se]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results