missing translation for 'onlineSavingsMsg'
Learn More

Copper AAS Standard, 1000 mg/L Cu in Ethanol 12%, HCl 2%, Honeywell Fluka™

Brand:  Honeywell Fluka E4201-500ML

Additional Details : Weight : 0.50000kg ??product.attributeSet.Transport?? : UN number : 2924 Chem class : 3 Pack group : III

 View more versions of this product

Product Code. 16654444

  • 115.46 € / 500mL
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.

Specifications

Specifications

Copper AAS Standard
1 g/cm3
<2
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions