missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLTA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | CLTA |
|---|---|
| Concentration | 0.47 mg/mL |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16877513
|
Proteintech
10852-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16867513
|
Proteintech
10852-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Clathrin consists of three heavy chain (192 kDa) and three light chains (LCa or LCb, 25-29 kDa) and self-assembles into a polyhedral cytoskeletal element. Self-assembly facilitates clathrin′s central role in membrane sorting and selective transport within the cell.Specifications
| CLTA | |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| O08585, P08081, P09496 | |
| 1211, 12757, 83800 | |
| CLTA Fusion Protein Ag1299 | |
| Primary | |
| -20°C | |
| CLTA |
| 0.47 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Clathrin light chain A, clathrin, light chain (Lca), CLTA, LCA | |
| Clta | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title