missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLTA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10852-1-AP-20UL
This item is not returnable.
View return policy
Description
Clathrin consists of three heavy chain (192 kDa) and three light chains (LCa or LCb, 25-29 kDa) and self-assembles into a polyhedral cytoskeletal element. Self-assembly facilitates clathrin′s central role in membrane sorting and selective transport within the cell.Specifications
| CLTA | |
| Polyclonal | |
| Unconjugated | |
| CLTA | |
| Clathrin light chain A, clathrin, light chain (Lca), CLTA, LCA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1211, 12757, 83800 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O08585, P08081, P09496 | |
| Clta | |
| CLTA Fusion Protein Ag1299 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction