Learn More
Clobenzorex Hydrochloride, Mikromol™
Description
Clobenzorex Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Clobenzorex Hydrochloride |
| CAS | 5843-53-8 |
| Analyte or Component Names | Clobenzorex Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H18 Cl N . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Clobenzorex Hydrochloride, Asenlix, Ba 7205, Dinintel, Rexigen, SD 271-12, Benzeneethanamine, N-[(2-chlorophenyl)methyl]-alpha-methyl-, hydrochloride (1:1), (+)-, Benzeneethanamine, N-[(2-chlorophenyl)methyl]-alpha-methyl-, hydrochloride, (+)- (9CI), Phenethylamine, N-(o-chlorobenzyl)-alpha-methyl-, hydrochloride, (+)- (8CI) |
| InChI Formula | InChI=1S/C16H18ClN.ClH/c1-13(11-14-7-3-2-4-8-14)18-12-15-9-5-6-10-16(15)17;/h2-10,13,18H,11-12H2,1H3;1H |
| SMILES | Cl.CC(Cc1ccccc1)NCc2ccccc2Cl |
| IUPAC Name | N-[(2-chlorophenyl)methyl]-1-phenylpropan-2-amine;hydrochloride |
| Show More |