Learn More
Cladribine EP Impurity C, Mikromol™
Description
Cladribine EP Impurity C
Specifications
Specifications
| Chemical Name or Material | 2-Chloroadenine |
| CAS | 1839-18-5 |
| Analyte or Component Names | 2-Chloroadenine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C5 H4 Cl N5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-Chloro-7H-purin-6-amine (2-Chloroadenine), 9H-Purin-6-amine, 2-chloro-, 1H-Purin-6-amine, 2-chloro- (9CI), Adenine, 2-chloro- (6CI,7CI,8CI), 2-Chloro-6-aminopurine, 2-Chloroadenine, 6-Amino-2-chloropurine, NSC 7362, SQ 22982, Cladribine Imp. C (EP), Cladribine Impurity C |
| InChI Formula | InChI=1S/C5H4ClN5/c6-5-10-3(7)2-4(11-5)9-1-8-2/h1H,(H3,7,8,9,10,11) |
| SMILES | Nc1nc(Cl)nc2nc[nH]c12 |
| IUPAC Name | 2-chloro-7H-purin-6-amine |
| Show More |