Learn More
cis-4-Methylcyclohexylamine Hydrochloride, Mikromol
Description
cis-4-Methylcyclohexylamine Hydrochloride, Mikromol
Specifications
Specifications
| Chemical Name or Material | cis-4-Methylcyclohexylamine Hydrochloride |
| CAS | 33483-66-8 |
| Analyte or Component Names | cis-4-Methylcyclohexylamine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H15 N . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | cis-4-Methylcyclohexylamine Hydrochloride |
| InChI Formula | InChI=1S/C7H15N.ClH/c1-6-2-4-7(8)5-3-6;/h6-7H,2-5,8H2,1H3;1H/t6-,7+; |
| Solution Type | Neat |
| SMILES | Cl.C[C@@H]1CC[C@H](N)CC1 |
| Show More |