Learn More
Cinnarizine N4-Oxide Dihydrochloride, Mikromol™
Description
Cinnarizine N4-Oxide Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | Cinnarizine N4-Oxide Dihydrochloride |
| CAS | 2734084-16-1 |
| Analyte or Component Names | Cinnarizine N4-Oxide Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H28 N2 O . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Cinnarizine N4-Oxide Dihydrochloride |
| InChI Formula | InChI=1S/C26H28N2O.2ClH/c29-28(20-10-13-23-11-4-1-5-12-23)21-18-27(19-22-28)26(24-14-6-2-7-15-24)25-16-8-3-9-17-25;;/h1-17,26H,18-22H2;2*1H/b13-10+;; |
| SMILES | Cl.Cl.[O-][N+]1(C\C=C\c2ccccc2)CCN(CC1)C(c3ccccc3)c4ccccc4 |
| IUPAC Name | 4-benzhydryl-1-oxido-1-[(E)-3-phenylprop-2-enyl]piperazin-1-ium;dihydrochloride |
| Show More |