Learn More
Chlorthiamid, LoGiCal
Product Code. 30457817
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 30457817
Brand: LoGiCal LGCFOR2572.00
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Chlorthiamid
CAS 1918-13-4
Analyte or Component Names Chlorthiamid
Shipping Condition Room Temperature
Molecular Formula C7 H5 Cl2 N S
Certifications/Compliance ISO 9001
Synonym Benzenecarbothioamide, 2,6-dichloro-, Benzamide, 2,6-dichlorothio- (7CI,8CI), 2,6-Dichlorothiobenzamide, Chlorthiamid, Chlorthiamide, DCBN, Prefix, Prefix (chlorthiamid), SD 7961
InChI Formula InChI=1S/C7H5Cl2NS/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H2,10,11)
Solution Type Neat
SMILES NC(=S)c1c(Cl)cccc1Cl
IUPAC Name 2,6-dichlorobenzenecarbothioamide
Industry Type Clinical Testing
Molecular Weight (g/mol) 206.09
Quantity 10 mg
Formula Weight 204.952 g/mol