Learn More
Chlorpromazine N-Oxide Hydrochloride, Mikromol™
Description
Chlorpromazine N-Oxide Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Chlorpromazine N-Oxide Hydrochloride |
| CAS | 2726144-94-9 |
| Analyte or Component Names | Chlorpromazine N-Oxide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H19 Cl N2 O S . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Chlorpromazine N-Oxide Hydrochloride |
| InChI Formula | InChI=1S/C17H19ClN2OS.ClH/c1-20(2,21)11-5-10-19-14-6-3-4-7-16(14)22-17-9-8-13(18)12-15(17)19;/h3-4,6-9,12H,5,10-11H2,1-2H3;1H |
| SMILES | Cl.C[N+](C)([O-])CCCN1c2ccccc2Sc3ccc(Cl)cc13 |
| IUPAC Name | 3-(2-chlorophenothiazin-10-yl)-N,N-dimethylpropan-1-amine oxide;hydrochloride |
| Show More |