missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Ceramus™ Fix Volume Bottle Top Dispensers

Product Code. 10702951
Change view
Click to view available options
Volume (Metric):
2 fix
25 fix
5 fix
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10702951 5 fix Each
10186050 2 fix Each
10277910 25 fix Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10702951 Supplier Hirschmann™ Supplier No. 9332100
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Guarantees consistent precision and volume dispensing with reproducible results independent of the user. The proven unit for manual dispensing, featuring superior-quality materials, simple handling and well-engineered user comfort to make laboratory routines easier.

  • For fixed and variable volumes, from 0.2 mL to 60 mL
  • Complete unit can be steam sterilized at 121°C without dismantling
  • Free rotation
  • Media recirculating system for simple and rapid venting without reagent loss
  • Digital volume configuration engagement, calibrated for a lifetime
  • Use within a temperature range of 4°C to 50°C
  • Individual quality certificate with conformity certificate

Specifications

Accuracy 0.6%
Product Type Bottletop Dispenser
Autoclavable Autoclavable
Certifications/Compliance ISO 8655
Description ceramus™ FIX, fixed volume bottle top dispenser with reagent recirculation system
Material Aluminum Oxide Ceramic Piston, Duran Glass Cylinder
Volume (Metric) 5 fix
For Use With (Equipment) Bottle
Includes Thread adapter, dispensing nozzle, suction tube, valve wrench, calibration report
Increments 0.05 mL
Precision 0.6%
Quantity 1 Ea.
Reproducibility 0.2%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.