Learn More
Manta Diagnostics™ Cellulose Nitrate Filters with Grid
200 filters per pack, individually wrapped
Brand: Manta Diagnostics™ AL1
Additional Details : Weight : 0.15000kg
Description
- Cellulose Nitrate
- 47 mm Diameter
- 200 per pack
- Individually wrapped
- Sterile
Specifications
Not Autoclavable | |
0.45 μm | |
Blue | |
200 | |
47 mm |
Cellulose Nitrate | |
Sterile | |
Yes | |
1 |