Learn More
STEMCELL Technologies CellAdhere™ Dilution Buffer
Product Code. 15547881
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15547881 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15547881 Supplier STEMCELL Technologies Supplier No. 07183
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Dilution buffer for matrix proteins
  • Vitronectin XF™, developed and manufactured by Nucleus Biologics, is an effective alternative to Corning™ Matrigel™
  • Vitronectin XF™ is a defined, xeno-free cell culture matrix that supports the growth and differentiation of human pluripotent stem cells
  • When used with mTeSR™1, mTeSR™ Plus, TeSR™-E8™, or TeSR™2, Vitronectin XF™ provides a defined culture system for the maintenance of human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells under feeder-free conditions
  • This system allows complete control over the culture environment, resulting in more consistent cell populations and reproducible results in downstream applications
  • Note: Catalog #07180 is the matrix only and requires additional reagents (Catalog #07183, 07174 or 05872, and 27147) available for purchase separately or in a kit with Vitronectin XF™ (Catalog #07190 or 07191)
Specifications
Description Endoderm, PSC-Derived; Pluripotent Stem Cells
Quantity 100 mL